Polyethylenglycol 8.000
Leverandør: G Biosciences
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
RC-078
RC-077
RC-078EA
1910
DKK
RC-078
RC-077
Polyethylenglycol 8.000
Polyethylenglycol 8.000
Formel:
H(OCH₂CH₂)nOH Molekylvægt: 8000 g/mol Kogepunkt: 250 Massefylde: 1.125 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01779615 CAS-nummer: 25322-68-3 EINECS: 500-038-2 Merck Index: 12,07729 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...