Biotin conjugation estimation, HOOK™ BiotinQuant
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BKC-01 BKC-03
BKC-01EA 3540 DKK
BKC-01 BKC-03
Biotin conjugation estimation, HOOK™ BiotinQuant
Proteinkoblings- og mærkningssystemer
HOOK™ BiotinQuant is designed for the estimation of biotin conjugation. It measures biotin using HABA [4’-hydroxyazobenzene-2-carboxylic acid] dye. HABA binds with avidin at the biotin-binding site.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78001acdffa396fac1198b2c9d735a293f43eecf&vid=4c23fbd12e32e29d864244e9981c37df79a2d46f&prid=web&clid=SAPDAM