General purpose gloves, AlphaTec® 87-305
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
87-305/10 87-305/7 87-305/8 87-305/9
ANSE87-305/10EA 0 DKK
ANSE87-305/10 ANSE87-305/7 ANSE87-305/8 ANSE87-305/9
General purpose gloves, AlphaTec® 87-305
Handsker
These hand-specific reusable gloves are made of 100% latex and feature a fish scale finish.
  • Unsupported
  • Liquid and mechanical resistant
  • Pinked cuffs
  • Chlorinated inside to minimise the risk of allergy
Ideal for applications in food processing (fish, meat, fruits and vegetables) or for general cleaning and maintenance work.
Thickness: 0,40 mm
Colour: Blue
EN 388 (X010), EN 374, Cat. III, AQL 0,65 Food contact approved
Certificeringer: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016, EN 388:2016 (X010X), Cat. III, AQL 0,65
Food contact approved