Recombinant streptavidin
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-584 786-585 786-586
786-584EA 2980 DKK
786-584 786-585 786-586
Recombinant streptavidin
Protein oprensnings Protein Oprensningsreagenser
Recombinant streptavidin from Streptomyces avidinii, produced in E.coli.
Streptavidin is a tetrameric protein that has no carbohydrate and thereby reduces the amount of non specific binding. The solubility of streptavidin (isoelectric pH 5) in aqueous buffer is lower than avidin, but the binding of streptavidin to biotin is similar to that of avidin.