Sulfhydryl coupling resin for antibody purification
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-794 786-795 786-796 786-806
786-794EA 3470 DKK
786-794 786-795 786-796 786-806
Sulfhydryl coupling resin for antibody purification
Protein oprensnings Protein Purification Resins
This sulphhydryl coupling resin is designed for coupling of peptides and proteins to a solid 6% agarose support through free sulfhydryl groups (-SH). The iodoacetyl groups of the sulfhydryl coupling resin specifically react with free sulfhydryls to form covalent, permanent thioether bonds.
  • Simple and efficient coupling
  • Reduced steric hindrance ensures greater binding of protein