Flow-through conductivity cells
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
301252 301254
663-1918EA 9660 DKK
663-1918 663-1919
Flow-through conductivity cells
Elektroder Elektroder til ledningsevnemåling
Four-electrode plastic flow-through measuring cells with low-polarisation graphite electrodes for applications in closed circuits.
  • Suitable for flow-through measurements in the lab
  • For medium and high conductivity ranges
  • Also ideal for testing special systems
For use with InoLab and ProfiLine meters.
Bestillingsoplysninger: Cable needs to be ordered separately.