RheBuild® kits for injection valves
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RE7125079 RE7725999
VARIRE7125079EA 0 DKK
VARIRE7125079 VARIRE7725999
RheBuild® kits for injection valves
Kromatografi forsyninger Kromatografi reservedele
RheBuild® kits containing genuine Rheodyne® parts are available for most Rheodyne® products. RheBuild® kits eliminate the need to order individual parts.
Leveringsinformation: Each RheBuild® kit includes all the parts, tools and instructions to maintain the precision performance of Rheodyne® valve platforms.