Pouring rings for laboratory bottles, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1089911 1089914 1089917
SCOT1089911EA 63.2 DKK
SCOT1089911 SCOT1089914 SCOT1089917
Pouring rings for laboratory bottles, DURAN®
Låg Flaskelåg
PP, coloured.
  • Size GL 45
  • Very good chemical resistance
  • Very good temperature resistance
For reagent and media bottles.