Cleanroom shoe covers, Kimtech™ A8
Leverandør: KIMBERLY CLARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39370 39372 39371
113-8198EA 1430 DKK
113-8198 113-8200 113-8199
Cleanroom shoe covers, Kimtech™ A8
Engangsbeklædning
These comfortable shoe covers are manufactured of white Spunbound PP with Unitrax™ PE film, making them ideal for use in clean manufacturing.
  • Seamless sole and elasticated toe and heel
  • Durable and waterproof
  • Low lint for contamination control
  • Latex- and silicone-free
  • Up to 4× more grip than leading alternative anti-slip covers
  • Three different sizes available to ensure comfortable and secure fit
  • Thread colour (blue, white or black) indicates size
Suitable for use in ISO 7 cleanrooms.
The superb traction provided by the shoe covers helps guard against injuries resulting from slips, skids and falls, which can be very common in working environments. They also provide wet and dry splash and particle protection, and can be recycled using Kimtech™ RightCycle programme.
Certificeringer: PPE Cat I (EU) 2016/425
Emballering: Double bagged.