Shoe covers, CPE
Leverandør: HUBEI ORIENT INTL TRADING
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-8134EA 1510 DKK
113-8134 113-8135 114-9047 114-9048 113-8129 114-9042 114-9037
Shoe covers, CPE
Engangsbeklædning
These disposable shoe covers are made of soft and durable CPE.
  • CPE is water resistant and provides good resistance against chemicals
  • Lightweight, breathable, dust- and waterproof
  • Elasticated at the ankle
  • Different packaging options available
Suitable for use in laboratories, hospitals, food processing, manufacturing and construction areas.
Bestil nu
Specifikationer
Environmentally Preferable
Køn
Outfit applikationer Laboratory,Food