Service Trolleys
Leverandør: TRESTON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
71007001 71007002 71007003 71007004 71007005 71007006 71007007
SOVE71007001EA 0 DKK
SOVE71007001 SOVE71007002 SOVE71007003 SOVE71007004 SOVE71007005 SOVE71007006 SOVE71007007
Service Trolleys
Vogne
The service trolley frame is equipped with a safety locking feature which allows only one drawer to be opened at a time and is covered with a a separate rubber mat that prevents items from slipping.
  • The two sides of the trolley have standard perforated panels which can be attached with brackets and mountings to suit required applications
Two fixed castors (Ø 150 mm)
Two swivel castors including brakes (Ø 100 mm)