Reusable earplugs, E-A-R™ Express™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EX01002 EX01001
111-1865EA 1260 DKK
111-1865 MMMAEX01001
Reusable earplugs, E-A-R™ Express™
Ørepropper Genanvendelige ørepropper
E-A-R™ Express™ Reusable Earplugs are recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can reduce noise levels by 28 dB, and have a hygienic insertion grip.
  • Easy to slide gently into ears with slow recovery foam
  • Foam tip gives a comfortable and effective seal
  • Features a hygienic insertion grip
  • Recognisable blue stem and yellow tip for easy identification
Protect hearing at work or during leisure activities with the E-A-R™ Express™ Reusable Earplugs. Designed to slide gently into the ear, the specially designed pod foam tip provides a comfortable and effective seal. The insertion grip eliminates the need to touch the foam, making them extremely hygienic. Get the best of both worlds with a patented slow recovery foam for a comfortable acoustic seal with quick and easy push-to-fit insertion.
Available in a recognisable yellow and blue for easy identification. Provides noise reduction to 28 dB SNR and fully tested and approved against EN 352-2. Model is compatible for use with the e 3M™ E-A-RFit™ Dual-Ear Validation System and available as a probed plug.
Certificeringer: EN 352-2
Advarsel: * One size fits all.