Bottles, clear, wide neck, with thread, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1160100 1160200 1160110 1160220
SCOT1160100EA 9710 DKK
SCOT1160100 SCOT1160200 SCOT1160110 SCOT1160220
Bottles, clear, wide neck, with thread, DURAN®
Flasker Glasflasker
DURAN®, borosilicate glass 3.3, clear, without screw cap and pouring ring.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These larger sized bottles/carboys are ideal for bulk storage and handling of both liquid and solid intermediates and final formulations.