Ag-AK, Immunofluorescence test, IMAGEN™
Leverandør: THERMO FISHER DIAGNOSTICS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S610830-2
S610930-2
S611030-2
S611130-2
S611230-2
S611330-2
S611430-6
OXOIS610830-2EA
738
DKK
OXOIS610830-2
OXOIS610930-2
OXOIS611030-2
OXOIS611130-2
OXOIS611230-2
OXOIS611330-2
OXOIS611430-6
Ag-AK, Immunofluorescence test, IMAGEN™
Mikrobiologiske testsystemer
Antigen-Antistof Test
Ag-AK, Immunoassay
IMAGEN™ is a qualitative direct immunofluorescence test used for detection and confirmation of the presence of virus in cell culture or in clinical specimen.
- Very specific