Slide mailers
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86.972 86.972.150
SARS86.972EA 0 DKK
SARS86.972 SARS86.972.150
Slide mailers
Transportbeholdere til objektglasholder
These mailing vessels are manufactured from polypropylene and is suitable for the safe transportation or storage of two glass microscope slides (or four if placed against each other).
  • The lid features a ventilation hole
Bestillingsoplysninger: Containers available with or without ventilation hole.
Emballering: 50 pieces per sachet.