ESD safety shoes, lace-up, uvex 1, 8543
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8543.8/50 8543.8/38 8543.8/39 8543.8/40 8543.8/41 8543.8/42 8543.8/43 8543.8/37 8543.8/36 8543.8/44 8543.8/45 8543.8/46 8543.8/47 8543.8/49 8543.8/35 8543.8/51 8543.8/52 8543.8/48 8543.9/35 8543.9/36 8543.9/37 8543.9/38 8543.9/39 8543.9/40 8543.9/41 8543.9/42 8543.9/43 8543.9/44 8543.9/45 8543.9/46 8543.9/47 8543.9/48 8543.9/49 8543.9/50 8543.9/51 8543.9/52
UVEX8543.8/50EA 930 DKK
UVEX8543.8/50 UVEX8543.8/38 UVEX8543.8/39 UVEX8543.8/40 UVEX8543.8/41 UVEX8543.8/42 UVEX8543.8/43 UVEX8543.8/37 UVEX8543.8/36 UVEX8543.8/44 UVEX8543.8/45 UVEX8543.8/46 UVEX8543.8/47 UVEX8543.8/49 UVEX8543.8/35 UVEX8543.8/51 UVEX8543.8/52 UVEX8543.8/48 UVEX8543.9/35 UVEX8543.9/36 UVEX8543.9/37 UVEX8543.9/38 UVEX8543.9/39 UVEX8543.9/40 UVEX8543.9/41 UVEX8543.9/42 UVEX8543.9/43 UVEX8543.9/44 UVEX8543.9/45 UVEX8543.9/46 UVEX8543.9/47 UVEX8543.9/48 UVEX8543.9/49 UVEX8543.9/50 UVEX8543.9/51 UVEX8543.9/52
ESD safety shoes, lace-up, uvex 1, 8543
Sko og støvler
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock-absorbent heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 11, 12
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC