ESD safety shoes, lace-up, uvex 1 ladies, 8562
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8562.8/36 8562.8/35 8562.8/39 8562.8/37 8562.8/38 8562.8/43 8562.8/42 8562.8/41 8562.8/40 8562.7/35 8562.7/36 8562.7/37 8562.7/38 8562.7/39 8562.7/40 8562.7/41 8562.7/42 8562.7/43
114-5812EA 974 DKK
114-5812 114-5568 114-5815 114-5813 114-5814 114-5819 114-5818 114-5817 114-5816 UVEX8562.7/35 UVEX8562.7/36 UVEX8562.7/37 UVEX8562.7/38 UVEX8562.7/39 UVEX8562.7/40 UVEX8562.7/41 UVEX8562.7/42 UVEX8562.7/43
ESD safety shoes, lace-up, uvex 1 ladies, 8562
Sko og støvler
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Grey/pink
Width: 11
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S2, SRC