Cell culture dishes, glass bottom, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
150680 150682
734-5089EA 1600 DKK
734-5089 734-5090
Cell culture dishes, glass bottom, Nunc™
Dyrkningsplader Celledyrkningsskåle
Borosilicate coverglass bottom, with lid. The Nunc™ glass bottom dish combines the convenience of a standard 35 mm cell culture dish with the imaging benefits of coverglass to provide the optimum optical characteristics required for high-magnification microscopy and confocal image analysis.
  • Optically clear and uniform surface, ideal for observation at high magnification
  • Dish lid minimises evaporation and keeps the contents free from airborne contaminants