Bottle top vacuum filtration systems, DigiFILTER™
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9308032 N9308034 N9308031
PERCN9308032EA 5280 DKK
PERCN9308032 PERCN9308034 PERCN9308031
Bottle top vacuum filtration systems, DigiFILTER™
Filtre Topfiltre til flasker
This vacuum assisted DigiFILTER™ assembly provides a quick and easy way to lter samples prior to analysis. Increase lab productivity by ltering particulates from digested samples in a matter of seconds directly from the digestion tube.