ESD safety shoes, lace-up, uvex 1 x-tended support, 8511
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8512.8/36 8511.8/41 8511.8/40 8511.8/39 8511.8/38 8511.8/37 8511.8/36 8511.8/35 8511.8/45 8511.8/50 8512.8/40 8512.8/39 8512.8/38 8512.8/37 8511.8/43 8512.8/35 8511.8/42 8511.8/49 8511.8/48 8511.8/47 8511.8/46 8511.8/44 8512.8/43 8512.8/50 8512.8/49 8512.8/48 8512.8/47 8512.8/46 8512.8/44 8512.8/42 8512.8/41 8512.8/45
UVEX8512.8/36EA 1080 DKK
UVEX8512.8/36 UVEX8511.8/41 UVEX8511.8/40 UVEX8511.8/39 UVEX8511.8/38 UVEX8511.8/37 UVEX8511.8/36 UVEX8511.8/35 UVEX8511.8/45 UVEX8511.8/50 UVEX8512.8/40 UVEX8512.8/39 UVEX8512.8/38 UVEX8512.8/37 UVEX8511.8/43 UVEX8512.8/35 UVEX8511.8/42 UVEX8511.8/49 UVEX8511.8/48 UVEX8511.8/47 UVEX8511.8/46 UVEX8511.8/44 UVEX8512.8/43 UVEX8512.8/50 UVEX8512.8/49 UVEX8512.8/48 UVEX8512.8/47 UVEX8512.8/46 UVEX8512.8/44 UVEX8512.8/42 UVEX8512.8/41 UVEX8512.8/45
ESD safety shoes, lace-up, uvex 1 x-tended support, 8511
Sko og støvler
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Side frame offers medial and lateral support for to protect against ankle injuries
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/silver
Width: 11
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC