ELISA chemiluminescent detection system, OptiBlaze™
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-302 786-539
786-302EA 7280 DKK
786-302 GENO786-539
ELISA chemiluminescent detection system, OptiBlaze™
Assays ELISA'er
OptiBlaze™ ELISA femto-HRP and femto-AP are stabilised ultra sensitive luminol and 1,2 dioxetane based horseradish peroxidase or alkaline phosphatase substrates for the detection of HRP or AP-conjugated antibodies during ELISA.
OptiBlaze™ kits are chemiluminescent detection systems for Enzyme-Linked Immunosorbent Assay (ELISA). The chemiluminescent substrates provided in the kit are ultra sensitive substrates developed for luminometer-based applications, specific for horseradish peroxidase or alkaline phosphatase labelled antibodies.