Disposal bins for serological pipettes, Bio-bin®
Leverandør: ECONIX LTD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BEXBBPIP6OR BEXBBPIP6YL BEXBBPIP6TS BEXBBPIP6 BEXBBPIP6PL
119-0227EA 1580 DKK
119-0227 119-0177 119-0228 119-0469 119-0226
Disposal bins for serological pipettes, Bio-bin®
Bokse Affaldsbeholdere
The 6 L Bio-bin® has been specifically designed to safely contain serological pipettes in labs.
  • Tall and thin: Best used with a stand
  • Fold-flat space efficient design, stackable pre- and post-use
  • Absorbent Bio-matt™ in the base of every bin can absorb 100 ml of fluid
  • Built-in lid and handles
  • Temporary and permanent seals
  • Fully autoclavable