Büchner vacuum filtration funnels
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VB30/08M VB40/02M VB40/08M VB60/02M VB60/06M VB60/08M VB95/04M VB95/08M VB40/06M VB60/04M VB95/06M
BIBBVB30/08MEA 4680 DKK
BIBBVB30/08M BIBBVB40/02M BIBBVB40/08M BIBBVB60/02M 511-0244 BIBBVB60/08M BIBBVB95/04M BIBBVB95/08M BIBBVB40/06M BIBBVB60/04M BIBBVB95/06M
Büchner vacuum filtration funnels
Tragte Filtreringstragte Büchner-tragte
Borosilicate glass 3.3
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Büchner vacuum filtration funnel with glass sintered disc. It is designed with ground glass cone to allow direct filtration into Büchner flasks.
Bestillingsoplysninger: Büchner flasks must be ordered separately.