Desiccator, vacuum, with plates
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5305-0609 5305-0910 5305-1212
NALG5305-0609EA 1960 DKK
NALG5305-0609 NALG5305-0910 467-3121
Desiccator, vacuum, with plates
Ekssikatorer Ekssikatorer, Plastik
Vacuum chamber systems include a transparent jar and neoprene gasketed vacuum plate with a tubing adapter that fits 6,4 mm Int.Ø tubing. Jars are available in polycarbonate (PC) or polyetherimide (PEI). White plates are made of polypropylene (PP) or polycarbonate (PC) and are supplied with neoprene gaskets.
  • Withstands a full vacuum without risk of implosion or shattering
  • Useful as a controlled environment chamber for degassing operations and freeze drying
Advarsel: Do not autoclave jars intended for vacuum service. Do not use with unsaturated halogenated hydrocarbons. Also, polycarbonate jars cannot be used with other organic solvents, acids or bases.