Baths and circulators, stainless steel baths
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8035100 8035200 8035500 8035600 10000414 10000417
IKAA8035100EA 19800 DKK
IKAA8035100 IKAA8035200 IKAA8035500 IKAA8035600 IKAA10000414 IKAA10000417
Baths and circulators, stainless steel baths
Cirkulatorsystemer Åbne cirkulatorsystemer Bade med aftagelige cirkulatorer
The ICC compact immersion circulators allow flexible coupling to water baths of different volumes. They are designed for use with liquids up to temperatures of 150 °C. The convenient carrying handle and the compact design allow easy transportation as well as comfortable use. An economic and attractive solution for basic applications such as sample tempering and heating glassware.
  • Adjustable safety circuit
  • Switch from internal to external temperature with one key (only for control unit)
  • Optical and acoustic warning, example: for over heating
  • Powerful pressure/suction pump