Chemical resistant gloves, UltraNeo 340
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34340010 34340018 34340019
MAPA34340010EA 80.5 DKK
MAPA34340010 MAPA34340018 MAPA34340019
Chemical resistant gloves, UltraNeo 340
Handsker
These industrial gloves made of neoprene/latex feature a textile lining for additional comfort.
  • Suitable for long-term wear
  • Great dexterity, due to the gloves reduced thickness
  • Scalloped cuff
  • Smooth exterior finish
Ideal for use in the automotive/mechanical industry, for batteries, pumps and compressor manufacturing, timber treatment, industrial cleaning, and for handling and transporting chemicals.
Thickness: 1,15 mm
Colour: Black
EN 374 (CKL), EN 388 (3121), EN 407 (X1XXXX), Cat. III; AQL 1,5
Certificeringer: EN ISO 374-1 Type A (CLMNST), EN ISO 374-5, EN 388:2016 (2121X), EN 407 (X1XXXX), Cat. III