Transport media, Ƹ Virocult®
Leverandør: MEDICAL WIRE EQUIPMENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MW951S MW950T50 MW951T
MEDWMW951SEA 3880 DKK
MEDWMW951S MEDWMW950T50 MEDWMW951T
Transport media, Ƹ Virocult®
Mikrobiologiske medier Klargjort medium
Ƹ Virocult tubes are used for respiratory or tissue sampling.
  • Optimum recovery of viruses at ambient and refrigerated conditions
  • Molecular and cell culture techniques
  • Antimicrobials prevent the growth of any bacteria and fungi
Compatible with molecular and cell culture system.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904.