Cartridge filters, Savana® AT
Leverandør: ENTEGRIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SVTA01TQ1 SVTB01TQ1 SVTV01TQ1 SVTC01TQ1 SVTG01TQ1
ENTESVTA01TQ1EA 0 DKK
ENTESVTA01TQ1 ENTESVTB01TQ1 ENTESVTV01TQ1 ENTESVTC01TQ1 ENTESVTG01TQ1
Cartridge filters, Savana® AT
Filtre Filterpatroner
These cartridge filters are designed with all fluoropolymer construction to consistently remove particles from a wide range of acids, bases and other process chemicals.
  • Excellent chemical compatibility
  • Manufactured in ISO 9001 certified facilities under cleanroom conditions and documentation control
Membrane: PTFE; Support: PFA
Maximum operating temperature: 120 °C
Maximum forward differential pressure: 5,1 bar at 25 °C; 2,0 bar at 120 °C
Maximum reverse differential pressure: 3,5 bar at 25 °C