Deflagrating spoon
Leverandør: R L ENTERPRISES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Z4704 Z4804
RLAAZ4704EA 86.4 DKK
RLAAZ4704 RLAAZ4804
Deflagrating spoon
Skeer
Mild steel. Deflagrating spoon, welded to a handle.
With cork and aluminium convex cap, 90 mm diameter to give safety closure.