Tubing, metal
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8930210 8930214 8930221 8930209 8930220 8930223 8930211 8930222
461-0005EA 3020 DKK
461-0005 461-0007 JULA8930221 461-0004 JULA8930220 JULA8930223 JULA8930211 JULA8930222
Tubing, metal
Slanger Overførselsslange
Metal tubing is integrated insulated and flexible with connectors and fittings. This flexible metal tubing can be bolted together with the circulator and the external system, provides anti-slipping. Triple insulated tubing is used for wide temperature ranges.