Jerricans, narrow neck, HDPE, UN Approved
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1428-0010 1428-0020 1428-0030
216-1955EA 724 DKK
216-1955 216-1956 216-1957
Jerricans, narrow neck, HDPE, UN Approved
Flasker Metaldunke
HDPE, black (eletrically conductive).
  • With carrying handle and screw cap
  • Good chemical resistance and stackable
  • With UN approval for transport
Electrically conductive canisters are particularly suitable for capable of diverting a charge and can be earthed, which makes them suitable for use with flammable liquids.