HPLC columns, MonoChrom® CN
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A0541050X020 A0541100X020 A0541250X046 A0506150X046 A0506250X046 A0509250X040 A0509250X046 A0509MG A0509MG2
VARIA0541050X020EA 0 DKK
VARIA0541050X020 VARIA0541100X020 VARIA0541250X046 VARIA0506150X046 VARIA0506250X046 VARIA0509250X040 VARIA0509250X046 VARIA0509MG VARIA0509MG2
HPLC columns, MonoChrom® CN
Kromatografikolonner
MonoChrom® columns provide excellent chromatography of organic amines, organic acids, amphoteric compounds and neutral compounds.
  • Excellent peak shape for basic analytes
  • High surface coverage
  • Improved polar compound retention
  • Enhanced selectivity