Spray bottles
Leverandør: LAMAPLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
394 395
215-4232EA 74.6 DKK
215-4232 LAMA395
Spray bottles
Flasker Sprayflasker
LDPE, translucent.
  • Good chemical resistance, good temperature resistance
  • Spray nozzle adjustment can be varied by rotation
  • Pressureless spray bottle
Spray, choice of fine mist or focused jet spray functions to disperse fluids.