Wafer Tweezers
Leverandør: OUTILS RUBIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41LB-4 42LB-6 41LB-8 41LB-6 41LB-5 43LB-10FT 43LB-14FT
OUTI41LB-4EA 377 DKK
OUTI41LB-4 OUTI42LB-6 OUTI41LB-8 OUTI41LB-6 OUTI41LB-5 OUTI43LB-10FT OUTI43LB-14FT
Wafer Tweezers
Pincet
Stainless steel. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddel for glare prevention
  • Each Wafer size has its corresponding tweezers size (example for an 8" Wafer take tweezers with a No. 8)