Prepared slides with platyhelminthes
Leverandør: LIEDER JOHANNES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PY313D PY2206E PY229G PY230E PY232I PY314D PY3154D PY3275E PY331G PY337F PY3392F PY3145F PY112C
LIEDPY313DEA 82.5 DKK
LIEDPY313D LIEDPY2206E LIEDPY229G LIEDPY230E LIEDPY232I LIEDPY314D LIEDPY3154D LIEDPY3275E LIEDPY331G LIEDPY337F LIEDPY3392F LIEDPY3145F LIEDPY112C
Prepared slides with platyhelminthes
Objektglas Prepared Slides
These prepared slides with platyhelminthes are designed for educational purposes.
  • Cestodes that is tapeworms infection of the digestive tract
  • Trematodes known as parasitic flatworms (flukes)
  • Turbellaria ie. planaria primitive traditional flatworms
(wm) whole mount (ts) transverse section (s) section.