HPLC columns, MetaSil AQ C18
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A0520150X020 A0530050G100
VARIA0520150X020EA 16780 DKK
VARIA0520150X020 VARIA0530050G100
HPLC columns, MetaSil AQ C18
Kromatografikolonner
The MetaSil AQ materials are based on silica that is highly coated with hydrophobic alkyl chains interspersed with hydrophilic short-chain endcapping groups. These columns remain fully extended in aqueous phases, allowing increased interaction between samples and the bonded phase. These columns remain fully extended in aqueous phases, allowing increased interaction between samples and the bonded phase.