Bag cutters
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BC-01IR BC-01
811-0165EA 7310 DKK
811-0165 MICRBC-01
Bag cutters
Knive
Plastic cutters for opening raw material packages.
  • Automatically retractable safety blade guard
  • No blade changes required - eliminates all loose blades
  • Ergonomic handle
  • Available gamma irradiated