Autoclavable systembox EasyMop GMP with MopJump-System
Leverandør: PFENNIG REINIGUNGSTECHNIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3500200 3500210 3500051 3500200QR
141-0952EA 5500 DKK
141-0952 141-0953 141-0951 141-1469
Autoclavable systembox EasyMop GMP with MopJump-System
Mopper
Autoclavable systembox EasyMop GMP and MopJump-System:
The newly developed 'EasyMop GMP' system box ensures compliance with regulations governing the cleaning and desinfecting of cleanrooms, and offers enormous advantages regarding its application.
  • All assembly components for use in pharmaceutical cleanrooms are certified
  • All measurements are precisely adapted to the intended use
  • The 'MopJump' system with displacement area allows easy impregnation of the mop head as well as a contact-free pick up
  • Triple-sealing ensures that the system box is securely closed
  • System box is suitable for 10 mops which clean and disinfect up to 100 m² floor Mop heads are autoclavable in the system box
  • The special construction allows the mops to be carried into the cleanroom
  • The system box can also be used as a stand-alone solution (ideal for small cleanrooms) or with a trolley (please contact Avantor for further information)
Mopp King GMP:
Applications: Cleaning and disinfection of GMP areas, up to 50 decontamination cycles. 'King GMP' is made of special yarn, especially suitable for cleanrooms, and provides maximum efficiency due to microfibres. This mop is especially developed for the impregnation method.
Safe disinfecting due to uniform moistening, the mop is washable, reusable and autoclavable
Can also be used for the traditional cleaning method with flat wringer
Also available with barcode and label (please contact your local Avantor sales office for further information)
Thermal stability: Melting point 250 to 260 °C
Absorption capacity: 3 ml/g
Fibre resistance (breaking strength): 14,0 kg
Elasticity: 57,5%
Material compatibility tested with the following products: Perform sterile concentrate OXY, perform classic concentrate OXY, Surfanios IP sterile, perform sterile concentrate QB, perform classic concentrate Q-Plus, perform classic concentrate QB, Buraton 10F, perform advanced concentrate QB.