Industrial gloves, Nitrile Knitwrist, A300
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A300NARL A300NARXL A300NARM
PRTWA300NARLEA 228 DKK
PRTWA300NARL PRTWA300NARXL PRTWA300NARM
Industrial gloves, Nitrile Knitwrist, A300
Handsker
These fully coated nitrile knitwrist gloves with jersey lining are designed for applications that require additional abrasion resistance.
  • Tough nitrile coating prevents grease, oil and water penetration
  • Cotton base for wearing comfort
Suitable for refuse collection, utilities and maintenance.
EN 388 (4111)
Certificeringer: EN 388:2016 (4111X).