SPE plates, 96-well, HyperSep™, Aminopropyl
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60300-505 60300-506 60300-507
HYPE60300-505EA 4380 DKK
HYPE60300-505 HYPE60300-506 HYPE60300-507
SPE plates, 96-well, HyperSep™, Aminopropyl
Mikroplader Solid Phase Ekstraktions plader
Thermo Scientific™ HyperSep Aminopropyl SPE 96-Well Plates and Individual Wells feature a polar sorbent that allows both polar and anion exchange interactions. The chemistry offers good retention of drugs, metabolites and structural isomers.
  • Separation of structural isomers, Saccharides, drugs and metabolites in biological fluids