Leather gloves, Stronghand® Welderstar
Leverandør: FELDTMANN HELMUT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0272/10 0272/8 0272/9
FELD0272/10EA 83.9 DKK
FELD0272/10 FELD0272/8 FELD0272/9
Leather gloves, Stronghand® Welderstar
Handsker
These gloves are made of cowhide nappa leather, the cuff consists of split leather.
  • Very soft and smooth leather
  • Sewn with aramid fibres
Ideal for Argon welding applications.
Colour: Beige/grey
EN 388; DIN EN 12477 exposition A
Certificeringer: EN 388:2016 (3131X); DIN EN 12477 exposition A