Glove liners, PU Fingertip, A121
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A121WHRL A121WHRXL A121WHRM A121WHRS A121WHRXS
PRTWA121WHRLEA 2910 DKK
PRTWA121WHRL PRTWA121WHRXL PRTWA121WHRM PRTWA121WHRS PRTWA121WHRXS
Glove liners, PU Fingertip, A121
Handsker
These knitted glove liners made of nylon are suitable for use in microelectronics, precision assembly and inspection.
  • Fingertips with dipped PU coating for protection against nicks and cuts
  • High abrasion resistance
Gauge: 13
Colour: White
Cat. I
Certificeringer: Cat. I