Adhesive PCR foil seals, QuickSeal Foil PCR ULTRA™
Leverandør: iST Scientific Ltd
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IST-129-080LR IST-129-080LS IST-129-080SR IST-129-080SS IST-129-080TS
731-0456EA 6680 DKK
731-0456 731-0457 731-0458 731-0459 731-0460
Adhesive PCR foil seals, QuickSeal Foil PCR ULTRA™
Forsegling Mikrotiterpladetætninger
Pierceable, peelable barrier foil for PCR and sample storage.
  • Seal integrity range: −70 to +120 °C
  • Available sterile or non sterile