Adhesive PCR film seals, QuickSeal PCR™
Leverandør: iST Scientific Ltd
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IST-120-080LR IST-120-080SR IST-120-080LS IST-120-080SS IST-120-080TS
731-0462EA 9700 DKK
731-0462 731-0448 731-0447 731-0449 731-0450
Adhesive PCR film seals, QuickSeal PCR™
Forsegling Mikrotiterpladetætninger
Transparent, peelable film for PCR.
  • Non pierceable
  • Available sterile or non sterile
  • Seal integrity range: −20 to +110 °C