Analytical balances, Adventurer
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30100620 30100622 30100621 30122610 30122657 30122611 30122612 30122613
611-3744EA 27590 DKK
611-3744 611-3791 611-3745 611-3213 611-3222 611-3214 611-3215 611-3216
Analytical balances, Adventurer
Vægte
Balances with multiple features, but uncomplicated weighing capabilities, provide the user with rapid, accurate results for routine weighing and measurement activities. Applications include weighing, formulation, check weighing and percentage weighing. Stabilisation times are typically less than three seconds.
  • Stability, accuracy and fast operation
  • Wide viewing angle colour touch screen (109 mm) for quick access to nine application modes
  • Dual-USB ports (front and rear) with GLP/GMP tracking
  • Two-piece, draft shield side doors provide good access and reduce footprint requirement when doors are open
  • Choice of balances with external calibration only or with AutoCal™ as additional feature
  • LIMS compatibility
Bestillingsoplysninger: For the full range of available accessories please contact Avantor.
Leveringsinformation: Supplied with AC adapter.
Advarsel: Model with suffix /E are external calibration only.