Glove liners, Puri Palm Fit
Leverandør: PURITECH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PFG500-S PFG500-M PFG500-L PFG500-XL PFG500-XXL PFG500-XS
112-4184EA 378 DKK
112-4184 112-4183 112-4182 112-4185 112-4187 112-4186
Glove liners, Puri Palm Fit
Handsker
These glove liners made of 100% soft nylon fabric with PU coating on the palm are ideal for handling small components.
  • Seamless liner
  • Hand-specific for great fit
  • Excellent dexterity and high absorbency
Can be used for light dry assembly, maintenance or micro mechanical tasks and for pharmaceutical processing.
Thickness of coating: 0,1 mm.