Immersion thermostats
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2000.0023.01 2061.0001.01
461-1047EA 15420 DKK
461-1047 HUBE2061.0001.01
Immersion thermostats
Cirkulatorsystemer Åbne cirkulatorsystemer Styreenheder til termostater
Immersion thermostats form the basis of many different combinations with polycarbonate or stainless steel bath vessels. All models are fitted with a powerful pressure/suction pump and meet the requirements of Safety Class III (FL) for use with flammable fluids. Temperatures down to −30 °C can be achieved in combination with a refrigerated bath. A screw clamp is included allowing the thermostats to be easily mounted on any vessels. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
  • Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
  • The controller can be upgraded with enhanced functionality at any time (Exclusive and Professional E-grade® packages), basic version features 11 languages (German, English, French, Italian, Spanish, Portuguese, Czech, Polish, Russian, Chinese and Japanese)
  • The temperature resolution can be displayed to 0,1 or 0,01 °C, display can be customised to the user's preferences and important information e.g. set point, actual internal or process temperatures can be displayed in a larger format
  • Powerful variable speed pressure/suction pump with variable pressure control (VPC)
  • User can set visual and acoustic alarms limiting the temperature range, and program the unit with its clock and calendar functions for automatic startup
Maximum pump capacities:
Pilot ONE® (CC-E): 27 L/min, 0,7 bar (pressure), 22 L/min, 0,4 bar suction
Pilot ONE® (CC-E xd): 22 L/min, 0,4 bar (pressure), 17 L/min, 0,25 bar suction
Modern and easy to use microprocessor controller with a large temperature display limited to essential functions only. MPC controller features over-temperature alarm and low liquid level protection. All MPC models also feature a powerful and suction pump for optimum performance.
Maximum pump capacities:
Pilot ONE® (CC-E): 27 l/min, 0,7 bar, 0,4 bar suction
Pilot ONE® (CC-E xd): 22 l/min, 0,4 bar, 0,4 bar suction
MPC-E: 20 l/min, 0,2 bar, 0,18 bar suction
Certificeringer: Safety Class III (FL) (DIN 12876) for use with flammable fluids
Bestillingsoplysninger: Accessories included. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
Advarsel: *Note: Working temperature range can be lowered to: −30 to +200 °C if an external cooling device is used.