Bottles, narrow neck, round, PS, with dip tube, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11681 11680 11650 11651 11655 11656 11666 11665
CORN11681EA 6900 DKK
CORN11681 CORN11680 CORN11650 CORN11651 CORN11655 CORN11656 CORN11666 CORN11665
Bottles, narrow neck, round, PS, with dip tube, Corning®
Flasker Plastflasker
Polystyrene, clear with HDPE screw cap.
  • Easy aseptic transfer of liquid
  • Good chemical resistance
  • Good temperature resistance
Sterile, double-bagged and preassembled. The preassembled transfer caps have two ports. One port ends in a 0,2 µm filter. The other port is heat sealable flexible tubing, ends in a male MPC or male luer lock connection.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904