Volumetric flasks, trapezoidal shape, class A, blue markings, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
936403 936404 936407 936406
BRND936403EA 653 DKK
BRND936403 BRND936404 BRND936407 BRND936406
Volumetric flasks, trapezoidal shape, class A, blue markings, BLAUBRAND®
Kolber Volumetriske kolber
Borosilicate glass 3,3 with PP or glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Individually certified, individual certificate included
* Wide neck
Certificeringer: DIN EN ISO 1042