Volumetric flasks, class A, blue markings
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2.37053 2.37052 2.37051 2.37049 2.37050
BRND2.37053EA 524 DKK
BRND2.37053 BRND2.37052 BRND2.37051 BRND2.37049 BRND2.37050
Volumetric flasks, class A, blue markings
Kolber Volumetriske kolber
Borosilicate glass 3.3, with beaded rim.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M mærkning
  • Total order from one batch, batch certificate included
Certificeringer: DIN EN ISO 1042